4-[(4-Chlorobenzyl)amino]benzoic acid

Supplier: Matrix Scientific

038396-500MG
101845-420EA 217.35 USD
101845-420
4-[(4-Chlorobenzyl)amino]benzoic acid
4-[(4-Chlorobenzyl)amino]benzoic acid

Hazard: Irritant

Formula: C₁₄H₁₂ClNO₂
MDL Number: MFCD01334921
CAS Number: 63759-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR