({[Benzyl(thien-2-ylmethyl)amino]carbonothioyl}amino)acetic acid

Supplier: Matrix Scientific

038390-500MG
101845-408EA 217.35 USD
101845-408
({[Benzyl(thien-2-ylmethyl)amino]carbonothioyl}amino)acetic acid
({[Benzyl(thien-2-ylmethyl)amino]carbonothioyl}amino)acetic acid

Hazard: Irritant

Formula: C₁₅H₁₆N₂O₂S₂
MDL Number: MFCD03450451
CAS Number: 656815-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR