[({(4-Methylbenzyl)[(5-methyl-2-furyl)methyl]amino}carbonothioyl)amino]acetic acid

Supplier: Matrix Scientific

038384-500MG
101845-396EA 217.35 USD
101845-396
[({(4-Methylbenzyl)[(5-methyl-2-furyl)methyl]amino}carbonothioyl)amino]acetic acid
[({(4-Methylbenzyl)[(5-methyl-2-furyl)methyl]amino}carbonothioyl)amino]acetic acid

Hazard: Irritant

Formula: C₁₇H₂₀N₂O₃S
MDL Number: MFCD03450522
CAS Number: 656815-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR