N-(3-Chlorophenyl)-N-propylamine

Supplier: Matrix Scientific

038374-500MG
101845-376EA 113.85 USD
101845-376
N-(3-Chlorophenyl)-N-propylamine
N-(3-Chlorophenyl)-N-propylamine

Hazard: Irritant

Formula: C₉H₁₂ClN
MDL Number: MFCD11144460
CAS Number: 31084-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR