2-Bromo-N-(2-ethyl-6-methylphenyl)butanamide

Supplier: Matrix Scientific

038364-500MG
101845-356EA 87.4 USD
101845-356
2-Bromo-N-(2-ethyl-6-methylphenyl)butanamide
2-Bromo-N-(2-ethyl-6-methylphenyl)butanamide

Hazard: Irritant

Formula: C₁₃H₁₈BrNO
MDL Number: MFCD03030403
CAS Number: 71394-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR