2-Bromo-N-(2-phenylethyl)butanamide

Supplier: Matrix Scientific

038362-500MG
101845-352EA 87.4 USD
101845-352
2-Bromo-N-(2-phenylethyl)butanamide
2-Bromo-N-(2-phenylethyl)butanamide

Hazard: Irritant

Formula: C₁₂H₁₆BrNO
MDL Number: MFCD01615489
CAS Number: 1119453-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR