2-Bromo-N-(3,4-dimethylphenyl)butanamide

Supplier: Matrix Scientific

038357-500MG
101845-342EA 87.4 USD
101845-342
2-Bromo-N-(3,4-dimethylphenyl)butanamide
2-Bromo-N-(3,4-dimethylphenyl)butanamide

Hazard: Irritant

Formula: C₁₂H₁₆BrNO
MDL Number: MFCD03030395
CAS Number: 1119451-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR