2-Bromo-N-(4-methoxyphenyl)butanamide

Supplier: Matrix Scientific

038347-500MG
101845-322EA 87.4 USD
101845-322
2-Bromo-N-(4-methoxyphenyl)butanamide
2-Bromo-N-(4-methoxyphenyl)butanamide

Hazard: Irritant

Formula: C₁₁H₁₄BrNO₂
MDL Number: MFCD02939927
CAS Number: 188491-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR