2-Bromo-N-(2-ethoxyphenyl)butanamide

Supplier: Matrix Scientific

038340-500MG
101845-308EA 87.4 USD
101845-308
2-Bromo-N-(2-ethoxyphenyl)butanamide
2-Bromo-N-(2-ethoxyphenyl)butanamide

Hazard: Irritant

Formula: C₁₂H₁₆BrNO₂
MDL Number: MFCD00297427
CAS Number: 1119450-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR