2-Bromo-N-(4-ethoxyphenyl)butanamide

Supplier: Matrix Scientific

038333-500MG
101845-294EA 87.4 USD
101845-294
2-Bromo-N-(4-ethoxyphenyl)butanamide
2-Bromo-N-(4-ethoxyphenyl)butanamide

Hazard: Irritant

Formula: C₁₂H₁₆BrNO₂
MDL Number: MFCD00297409
CAS Number: 451460-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR