2-Bromo-N-cyclopentylbutanamide

Supplier: Matrix Scientific

038331-500MG
101845-290EA 87.4 USD
101845-290
2-Bromo-N-cyclopentylbutanamide
2-Bromo-N-cyclopentylbutanamide

Hazard: Irritant

Formula: C₉H₁₆BrNO
MDL Number: MFCD03030361
CAS Number: 905811-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR