Dimethyl-2-isocyanatoterephthalate

Supplier: Matrix Scientific

038316-500MG
101845-258EA 144.9 USD
101845-258
Dimethyl-2-isocyanatoterephthalate
Dimethyl-2-isocyanatoterephthalate

Hazard: Irritant

Formula: C₁₁H₉NO₅
MDL Number: MFCD10699354
CAS Number: 179114-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR