4-Isocyanato-1,2-dimethoxybenzene

Supplier: Matrix Scientific

038309-500MG
101845-236EA 125.35 USD
101845-236
4-Isocyanato-1,2-dimethoxybenzene
4-Isocyanato-1,2-dimethoxybenzene

Hazard: Irritant

Formula: C₉H₉NO₃
MDL Number: MFCD01863688
CAS Number: 37527-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR