3-(4-Butoxyphenyl)-1H-pyrazole-4-carboxylic acid

Supplier: Matrix Scientific

038302-500MG
101845-224EA 217.35 USD
101845-224
3-(4-Butoxyphenyl)-1H-pyrazole-4-carboxylic acid
3-(4-Butoxyphenyl)-1H-pyrazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₃
MDL Number: MFCD12027306
CAS Number: 879996-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR