Ethyl [(4-methylphenyl)amino]acetate

Supplier: Matrix Scientific

038294-500MG
101845-208EA 109.25 USD
101845-208
Ethyl [(4-methylphenyl)amino]acetate
Ethyl [(4-methylphenyl)amino]acetate

Hazard: Irritant

Formula: C₁₁H₁₅NO₂
MDL Number: MFCD11154309
CAS Number: 21911-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR