1-(2-Chloroethyl)-4-methylpiperazine hydrochloride

Supplier: Matrix Scientific

038290-500MG
101845-200EA 39.1 USD
101845-200
1-(2-Chloroethyl)-4-methylpiperazine hydrochloride
1-(2-Chloroethyl)-4-methylpiperazine hydrochloride

Hazard: Irritant

Formula: C₇H₁₅ClN₂·ClH
MW: 199.121 g/mol
MDL Number: MFCD12031872
CAS Number: 5753-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR