1-[4-(4-Methylpiperidin-1-yl)phenyl]methanamine

Supplier: Matrix Scientific

038277-500MG
101845-174EA 217.35 USD
101845-174
1-[4-(4-Methylpiperidin-1-yl)phenyl]methanamine
1-[4-(4-Methylpiperidin-1-yl)phenyl]methanamine

Hazard: Irritant

Formula: C₁₃H₂₀N₂
MDL Number: MFCD04971042
CAS Number: 486437-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR