1-Ethyl-4-nitro-1H-pyrazole-3-carboxylic acid

Supplier: Matrix Scientific

038259-500MG
101845-138EA 217.35 USD
101845-138
1-Ethyl-4-nitro-1H-pyrazole-3-carboxylic acid
1-Ethyl-4-nitro-1H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₆H₇N₃O₄
MDL Number: MFCD02090836
CAS Number: 401906-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR