5-Methyl-4-[(3-methylphenoxy)methyl]isoxazole-3-carboxylic acid

Supplier: Matrix Scientific

038237-500MG
101845-092EA 289.8 USD
101845-092
5-Methyl-4-[(3-methylphenoxy)methyl]isoxazole-3-carboxylic acid
5-Methyl-4-[(3-methylphenoxy)methyl]isoxazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₃NO₄
MDL Number: MFCD06805570
CAS Number: 905809-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR