5-Bromoindole-2-carboxylic acid
Supplier: Matrix Scientific
Synonyms:
5-Bromo-1H-indole-2-carboxylic acid
038225-25G
038225-1G
101845-068EA
412.31
USD
101845-068
101845-064
5-Bromoindole-2-carboxylic acid
5-Bromoindole-2-carboxylic acid
Hazard: Irritant
Formula:
C₉H₆BrNO₂ MW: 240.06 g/mol |
MDL Number:
MFCD00022705 CAS Number: 7254-19-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...