2-(Methylamino)-N-propylacetamide

Supplier: Matrix Scientific

038208-500MG
101845-030EA 144.9 USD
101845-030
2-(Methylamino)-N-propylacetamide
2-(Methylamino)-N-propylacetamide

Hazard: Irritant

Formula: C₆H₁₄N₂O
MDL Number: MFCD05270871
CAS Number: 901273-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR