N-Isopropyl-2-(methylamino)acetamide

Supplier: Matrix Scientific

038204-500MG
101845-022EA 165.6 USD
101845-022
N-Isopropyl-2-(methylamino)acetamide
N-Isopropyl-2-(methylamino)acetamide

Hazard: Irritant

Formula: C₆H₁₄N₂O
MDL Number: MFCD06655723
CAS Number: 51489-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR