5-Chlorooxindole

Supplier: Matrix Scientific

Synonyms: 5-Chloro-2-oxindole

038190-500MG
101844-994EA 290.77 USD
101844-994
5-Chlorooxindole
5-Chlorooxindole

Hazard: Irritant

Formula: C₈H₆ClNO
MW: 167.59 g/mol
MDL Number: MFCD00191927
CAS Number: 17630-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR