1-Methyl-4-(piperazin-1-ylacetyl)piperazine

Supplier: Matrix Scientific

038180-500MG
101844-974EA 217.35 USD
101844-974
1-Methyl-4-(piperazin-1-ylacetyl)piperazine
1-Methyl-4-(piperazin-1-ylacetyl)piperazine

Hazard: Irritant

Formula: C₁₁H₂₂N₄O
MDL Number: MFCD08741847
CAS Number: 94012-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR