2-(4-Methoxyphenoxy)ethanimidamide

Supplier: Matrix Scientific

038178-500MG
101844-970EA 144.9 USD
101844-970
2-(4-Methoxyphenoxy)ethanimidamide
2-(4-Methoxyphenoxy)ethanimidamide

Hazard: Irritant

Formula: C₉H₁₂N₂O₂
MDL Number: MFCD08059925
CAS Number: 133915-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR