(1-Acetylpiperidin-4-yl)acetic acid

Supplier: Matrix Scientific

038174-500MG
101844-962EA 146.05 USD
101844-962
(1-Acetylpiperidin-4-yl)acetic acid
(1-Acetylpiperidin-4-yl)acetic acid

Hazard: Irritant

Formula: C₉H₁₅NO₃
MW: 185.22 g/mol
MDL Number: MFCD12027293
CAS Number: 885274-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR