2-Piperazin-1-yl-N-propylacetamide

Supplier: Matrix Scientific

038170-500MG
101844-954EA 289.8 USD
101844-954
2-Piperazin-1-yl-N-propylacetamide
2-Piperazin-1-yl-N-propylacetamide

Hazard: Irritant

Formula: C₉H₁₉N₃O
MDL Number: MFCD06655510
CAS Number: 39890-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR