4-(3-Methylphenyl)piperidin-4-ol

Supplier: Matrix Scientific

038130-500MG
101844-874EA 289.8 USD
101844-874
4-(3-Methylphenyl)piperidin-4-ol
4-(3-Methylphenyl)piperidin-4-ol

Hazard: Irritant

Formula: C₁₂H₁₇NO
MDL Number: MFCD08689267
CAS Number: 71916-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR