5-(2-Methoxyphenyl)cyclohexane-1,3-dione

Supplier: Matrix Scientific

038114-500MG
101844-842EA 217.35 USD
101844-842
5-(2-Methoxyphenyl)cyclohexane-1,3-dione
5-(2-Methoxyphenyl)cyclohexane-1,3-dione

Hazard: Irritant

Formula: C₁₃H₁₄O₃
MDL Number: MFCD03844402
CAS Number: 55579-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR