5-(2-Chlorophenyl)cyclohexane-1,3-dione

Supplier: Matrix Scientific

038097-500MG
101844-808EA 235.75 USD
101844-808
5-(2-Chlorophenyl)cyclohexane-1,3-dione
5-(2-Chlorophenyl)cyclohexane-1,3-dione

Hazard: Irritant

Formula: C₁₂H₁₁ClO₂
MDL Number: MFCD02067868
CAS Number: 55579-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR