3-Methyl-4-phenyl-1H-pyrazole

Supplier: Matrix Scientific

038095-500MG
101844-804EA 217.35 USD
101844-804
3-Methyl-4-phenyl-1H-pyrazole
3-Methyl-4-phenyl-1H-pyrazole

Hazard: Irritant

Formula: C₁₀H₁₀N₂
MDL Number: MFCD00051752
CAS Number: 13788-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR