3-[3-(Ethoxycarbonyl)-2,5-dimethyl-1H-pyrrol-1-yl]propanoic acid

Supplier: Matrix Scientific

038089-500MG
101844-792EA 162.15 USD
101844-792
3-[3-(Ethoxycarbonyl)-2,5-dimethyl-1H-pyrrol-1-yl]propanoic acid
3-[3-(Ethoxycarbonyl)-2,5-dimethyl-1H-pyrrol-1-yl]propanoic acid

Hazard: Irritant

Formula: C₁₂H₁₇NO₄
Melting Pt: 90 °C
MDL Number: MFCD09871352
CAS Number: 876294-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR