(1,3,5-Trimethyl-1H-pyrazol-4-yl)acetic acid

Supplier: Matrix Scientific

033622-500MG
101836-048EA 328.9 USD
101836-048
(1,3,5-Trimethyl-1H-pyrazol-4-yl)acetic acid
(1,3,5-Trimethyl-1H-pyrazol-4-yl)acetic acid

Hazard: Irritant

Formula: C₈H₁₂N₂O₂
MDL Number: MFCD08444355
CAS Number: 70598-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR