1-Phenyl-1,2,3,4-tetrahydropyrrolo[1,2-a]pyrazine

Supplier: Matrix Scientific

033595-500MG
101835-992EA 58.65 USD
101835-992
1-Phenyl-1,2,3,4-tetrahydropyrrolo[1,2-a]pyrazine
1-Phenyl-1,2,3,4-tetrahydropyrrolo[1,2-a]pyrazine

Hazard: Irritant

Formula: C₁₃H₁₄N₂
CAS Number: 112758-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR