2-Methylpyrimidine-5-carbaldehyde

Supplier: Matrix Scientific

033552-500MG
101835-906EA 43.7 USD
101835-906
2-Methylpyrimidine-5-carbaldehyde
2-Methylpyrimidine-5-carbaldehyde

Hazard: Irritant

Formula: C₆H₆N₂O
MW: 122.1246 g/mol
MDL Number: MFCD07186434
CAS Number: 90905-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR