2-(4-Methylpiperidine-1-sulfonyl)ethylamine oxalate

Supplier: Matrix Scientific

033551-500MG
101835-904EA 230 USD
101835-904
2-(4-Methylpiperidine-1-sulfonyl)ethylamine oxalate
2-(4-Methylpiperidine-1-sulfonyl)ethylamine oxalate

Hazard: Irritant

Formula: C₁₀H₂₀N₂O₆S
MDL Number: MFCD11506529
CAS Number: 924873-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR