3-(Methylpyridin-2-ylmethylamino)propionic acid dihydrochloride

Supplier: Matrix Scientific

033547-500MG
101835-896EA 230 USD
101835-896
3-(Methylpyridin-2-ylmethylamino)propionic acid dihydrochloride
3-(Methylpyridin-2-ylmethylamino)propionic acid dihydrochloride

Hazard: Irritant

Formula: C₁₀H₁₆Cl₂N₂O₂
MDL Number: MFCD11506527
CAS Number: 1185304-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR