Dimethylamino(3-fluorophenyl)acetic acid

Supplier: Matrix Scientific

033529-500MG
101835-860EA 230 USD
101835-860
Dimethylamino(3-fluorophenyl)acetic acid
Dimethylamino(3-fluorophenyl)acetic acid

Hazard: Irritant

Formula: C₁₀H₁₂FNO₂
MDL Number: MFCD08059912
CAS Number: 1007878-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR