(2-Ethylimidazol-1-yl)acetic acid hydrochloride

Supplier: Matrix Scientific

033524-500MG
101835-850EA 230 USD
101835-850
(2-Ethylimidazol-1-yl)acetic acid hydrochloride
(2-Ethylimidazol-1-yl)acetic acid hydrochloride

Hazard: Irritant

Formula: C₇H₁₁ClN₂O₂
MDL Number: MFCD11506517
CAS Number: 1170001-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR