(3-Ethylisoxazol-5-yl)methanol

Supplier: Matrix Scientific

033513-500MG
101835-828EA 184 USD
101835-828
(3-Ethylisoxazol-5-yl)methanol
(3-Ethylisoxazol-5-yl)methanol

Hazard: Irritant

Formula: C₆H₉NO₂
MDL Number: MFCD11048458
CAS Number: 14716-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR