3-Isobutylisoxazole-5-carboxylic acid

Supplier: Matrix Scientific

033512-500MG
101835-826EA 307.69 USD
101835-826
3-Isobutylisoxazole-5-carboxylic acid
3-Isobutylisoxazole-5-carboxylic acid

Hazard: Irritant

Formula: C₈H₁₁NO₃
MDL Number: MFCD08059886
CAS Number: 910321-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR