(1-Ethylpiperidin-4-yl)acetic acid

Supplier: Matrix Scientific

033502-500MG
101835-806EA 188.6 USD
101835-806
(1-Ethylpiperidin-4-yl)acetic acid
(1-Ethylpiperidin-4-yl)acetic acid

Hazard: Irritant

Formula: C₉H₁₇NO₂
MDL Number: MFCD08059874
CAS Number: 915922-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR