3-Piperidin-1-ylbenzylamine

Supplier: Matrix Scientific

033484-500MG
101835-770EA 230 USD
101835-770
3-Piperidin-1-ylbenzylamine
3-Piperidin-1-ylbenzylamine

Hazard: Irritant

Formula: C₁₂H₁₈N₂
MW: 190.29 g/mol
MDL Number: MFCD06659082
CAS Number: 175696-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR