2-(1-Methylpiperidin-2-ylmethoxy)phenylamine

Supplier: Matrix Scientific

033471-500MG
101835-744EA 230 USD
101835-744
2-(1-Methylpiperidin-2-ylmethoxy)phenylamine
2-(1-Methylpiperidin-2-ylmethoxy)phenylamine

Hazard: Irritant

Formula: C₁₃H₂₀N₂O
MDL Number: MFCD04970978
CAS Number: 869945-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR