4-(1-Pyrrolidinyl)aniline

Supplier: Matrix Scientific

Synonyms: 4-(Pyrrolidin-1-yl)aniline

033452-500MG
101835-706EA 238.46 USD
101835-706
4-(1-Pyrrolidinyl)aniline
4-(1-Pyrrolidinyl)aniline

Hazard: Irritant

Formula: C₁₀H₁₄N₂
MW: 162.2316 g/mol
MDL Number: MFCD00156245
CAS Number: 2632-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR