1-(4-Fluorobenzyl)piperidine-2-carboxylic acid

Supplier: Matrix Scientific

033445-500MG
101835-694EA 230 USD
101835-694
1-(4-Fluorobenzyl)piperidine-2-carboxylic acid
1-(4-Fluorobenzyl)piperidine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₆FNO₂
MW: 237.28 g/mol
MDL Number: MFCD05174530
CAS Number: 1030610-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR