2-(5-Methylthiophen-2-yl)ethylamine

Supplier: Matrix Scientific

033432-500MG
101835-668EA 230 USD
101835-668
2-(5-Methylthiophen-2-yl)ethylamine
2-(5-Methylthiophen-2-yl)ethylamine

Hazard: Irritant

Formula: C₇H₁₁NS
MW: 141.24 g/mol
MDL Number: MFCD08059759
CAS Number: 30433-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR