2-Benzyl-1,3-thiazole-4-carboxylic acid

Supplier: Matrix Scientific

033430-500MG
101835-664EA 134.55 USD
101835-664
2-Benzyl-1,3-thiazole-4-carboxylic acid
2-Benzyl-1,3-thiazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₉NO₂S
MDL Number: MFCD02102513
CAS Number: 36916-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR