4,6-Dimethyl-2-piperazin-1-ylpyrimidine

Supplier: Matrix Scientific

033417-500MG
101835-638EA 230 USD
101835-638
4,6-Dimethyl-2-piperazin-1-ylpyrimidine
4,6-Dimethyl-2-piperazin-1-ylpyrimidine

Hazard: Irritant

Formula: C₁₀H₁₆N₄
MW: 192.27 g/mol
MDL Number: MFCD00885959
CAS Number: 22746-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR