(4-Methylfurazan-3-ylmethylsulfanyl)acetic acid

Supplier: Matrix Scientific

033407-500MG
101835-618EA 230 USD
101835-618
(4-Methylfurazan-3-ylmethylsulfanyl)acetic acid
(4-Methylfurazan-3-ylmethylsulfanyl)acetic acid

Hazard: Irritant

Formula: C₆H₈N₂O₃S
MDL Number: MFCD07801110
CAS Number: 876715-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR